3,5-Xylenol ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
5-Hydroxy-3-xylene, 3,5-Dimethylphenol, 5-Hydroxy-m-xylene, 1,3-Dimethyl-5-hydroxybenzene
D0778-500G
D0778-25G
TCD0778-500GEA
647
USD
TCD0778-500G
TCD0778-25G
3,5-Xylenol ≥99.0%
3,5-Xylenol
CAS Number: 108-68-9
MDL Number: MFCD00002307
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 64
Flash Point (°C): 107
Other Product Names / Synonyms: m-Xylenol
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 220 °C Melting Pt: 64 °C Density: 0.968 g/cm³ (20 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002307 CAS Number: 108-68-9 EINECS: 203-606-5 UN: 2261 ADR: 6.1,II Merck Index: 12,10215 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...