High Fidelity DNA Polymerase
Supplier: G-Biosciences
786-1966 786-1967
76716-668EA 94.4 USD
76716-668 76716-630
High Fidelity DNA Polymerase
Nucleic Acid Reagents End-point PCR Enzymes and Kits
High-fidelity DNA Polymerase possesses 5´→3´ polymerase and 3´→5´ exonuclease (proofreading) activities. This Polymerase has the fidelity, sensitivity and processivity with an error rate ~2.8×10² fold lower than Taq DNA polymerase, and significantly lower than other proofreading DNA polymerases.
  • High fidelity PCR and cloning
Long and high GC content template DNA amplification.