Tetrabromomethane ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Carbon tetrabromide
T0038-500G
T0038-25G
T0038-100G
TCT0038-500GEA
270.59
USD
TCT0038-500G
TCT0038-25G
TCT0038-100G
Tetrabromomethane ≥99.0%
Tetrabromomethane
CAS Number: 558-13-4
MDL Number: MFCD00000117
Molecular Formula: CBr4
Molecular Weight: 331.63
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 190
Melting point (°C): 94
Product Categories: Synthetic Organic Chemistry, Carbon Homologation / Degradation Reaction, Halogenation
Other Product Names / Synonyms: Tetrabromomethane
Formula:
Br₄C MW: 331.63 g/mol Boiling Pt: 190 °C Melting Pt: 94 °C Density: 3.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000117 CAS Number: 558-13-4 EINECS: 209-189-6 UN: 2516 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...