cis-1,2-Diaminocyclohexane ≥97.0%

Supplier: TCI America

Synonyms: cis-Cyclohexane-1,2-diamine, (±)-cis-1,2-Diaminocyclohexane

C1463-5G C1463-25G
TCC1463-005GEA 77.23 USD
TCC1463-005G TCC1463-25G
cis-1,2-Diaminocyclohexane ≥97.0%
cis-1,2-Diaminocyclohexane

CAS Number: 1436-59-5
MDL Number: MFCD00063746
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 71
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: cis-1,2-Diaminocyclohexane

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 93 °C
Density: 0.97 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00063746
CAS Number: 1436-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ebc960d70a59896f9d36fb168aab42f4988b3c5&vid=4ebc960d70a59896f9d36fb168aab42f4988b3c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR