(2-Chlorophenyl)acetonitrile ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Chlorobenzyl cyanide, 2-(2-Chlorophenyl)acetonitrile

C1125-500G C1125-25G
TCC1125-500GEA 262 USD
TCC1125-500G TCC1125-25G
(2-Chlorophenyl)acetonitrile ≥98.0%
(2-Chlorophenyl)acetonitrile

CAS Number: 2856-63-5
MDL Number: MFCD00001898
Molecular Formula: C8H6ClN
Molecular Weight: 151.59
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 141
Freezing point (°C): 23


Other Product Names / Synonyms: (2-Chlorophenyl)acetonitrile

Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 21…24 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00001898
CAS Number: 2856-63-5
EINECS: 220-669-4
UN: 3439
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e116c3972810c18f297b7bf3d1dd6941e641e1fa&vid=e116c3972810c18f297b7bf3d1dd6941e641e1fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR