Clofibrate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate

C0941-25G C0941-500G
TCC0941-025GEA 70.52 USD
TCC0941-025G TCC0941-500G
Clofibrate ≥98.0%
Clofibrate

CAS Number: 637-07-0
MDL Number: MFCD00000615
Molecular Formula: C12H15ClO3
Molecular Weight: 242.70
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 1.14


Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2-(4-Chlorophenoxy)isobutyric Acid Ethyl Ester, Ethyl 2-(4-Chlorophenoxy)isobutyrate

Formula: C₁₂H₁₅ClO₃
Boiling Pt: 150 °C
Density: 1.14 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000615
CAS Number: 637-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97774f6a3556db923bd89fe20781154c2ccf9c1f&vid=97774f6a3556db923bd89fe20781154c2ccf9c1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR