Chlorosulfonyl isocyanate ≥98.0%

Supplier: TCI America

C0886-25G C0886-500G C0886-100G
TCC0886-025GEA 40.24 USD
TCC0886-025G TCC0886-500G TCC0886-100G
Chlorosulfonyl isocyanate ≥98.0%
Chlorosulphonyl isocyanate

CAS Number: 1189-71-5
MDL Number: MFCD00011608
Molecular Formula: CClNO3S
Molecular Weight: 141.53
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Melting point (°C): -80
Flash Point (°C): 110
Specific Gravity (20/20): 1.64
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: CSI

Formula: CClNO₃S
MW: 141.53 g/mol
Boiling Pt: 107 °C
Melting Pt: -80 °C
Density: 1.64 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00011608
CAS Number: 1189-71-5
EINECS: 214-715-2
UN: 3264
ADR: 8,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=746cffb0636acb6236846bedeb010c1bee508e89&vid=746cffb0636acb6236846bedeb010c1bee508e89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR