2-Chloronaphthalene ≥98.0%
Supplier: TCI America
C0213-1G
C0213-25G
TCC0213-001GEA
41
USD
TCC0213-001G
TCC0213-025G
2-Chloronaphthalene ≥98.0%
2-Chloronaphthalene
CAS Number: 91-58-7
MDL Number: MFCD00035731
Molecular Formula: C10H7Cl
Molecular Weight: 162.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 256
Melting point (°C): 59
Flash Point (°C): 125
Formula:
C₁₀H₇Cl Boiling Pt: 256 °C Melting Pt: 59 °C Density: 1.2656 g/cm³ (16 °C) Flash Pt: 125 °C |
MDL Number:
MFCD00035731 CAS Number: 91-58-7 EINECS: 202-079-9 Merck Index: 13,02169 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...