Nitrilite® 93-311, Nitrile Gloves, Ansell
Supplier: ANSELL COMMERCIAL MEXICO SA DE MX
365754 365755 365753 365756 365752
47748-214CS 626.13 USD
47748-214 47748-216 47748-212 47748-218 47748-210
Nitrilite® 93-311, Nitrile Gloves, Ansell
Gloves
Specifically designed for strength and comfort in critical environments, Nitrilite 93-311 significantly reduces particulate and extractable levels.
  • Strength and comfort in critical environments
  • Significantly reduces particulate and extractable levels
It is 100% powder-free with no plasicizers, silicone, or pigments.
Ideal for electronics manufacturing, medical laboratories, pharmaceutical, and research laboratories.
Its nitrile material is thinner than comparable vinyl or latex gloves for a lighter, more comfortable glove with improved dexterity and tactility. Ideal applications include electronics assembly, handling sensitive parts and materials, pharmaceutical manufacturing, product protection in handling: cosmetics packaging , optical goods , microelectronic and micro-mechanic components, and protection from blood and other bodily fluids.
Certificaciones: Manufactured in ISO 9002 registered facilities. Each lot is tested, certified, and documented. AQL for pinholes: 4.0.
Envase: Gloves feature cleanroom-compatible packaging containing 50 gloves per inner poly bag and 2 inner bags per outer poly bag. Lot number is printed on outer poly bag in IPA-resistant ink.