AlphaTec® 88-394 Natural Rubber Latex Gloves, Unlined, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
120129 120130 120131 120132
32890-365CS 454.18 USD
32890-365 32890-366 32890-368 32890-369
AlphaTec® 88-394 Natural Rubber Latex Gloves, Unlined, Ansell
Gloves
Gloves provide excellent dexterity, tensile strength and elasticity, along with chemical, snag, puncture and abrasion resistance.
  • Excellent dexterity and tensile strength
  • Strong chemical and puncture resistance
  • Ideal glove for safety and dexterity in the food processing industry
Use in the agriculture, viticulture, food processing and food service industries. Uses include canning and food processing, food packing and handling, fruit and vegetable processing, light-duty maintenance and clean-up, and general industry.
Certificaciones: Complies with FDA food handling requirements (21 CFR 177.2600).
Envase: Gloves are packaged 12 pairs in a bag; 144 in a carton.