(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulfonate) ≥98%, white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO
CHAPSO is the hydroxy analog of CHAPS; a non-denaturing zwitterionic detergent for solubilizing proteins, especially integral membrane proteins while maintaining the functional capability of the protein of interest.
- Useful for the solubilization of integral membrane proteins
- Critical micelle concentration (CMC): 8 mM
- Aggregation Number: 11
It has a more polar head group than CHAPS, giving it a greater solubility. CHAPSO has a high CMC (8 mM) and does not denature membrane-bound proteins. It can solubilise the opiate receptor in a state exhibiting reversible binding of opiates.
A nondenaturing zwitterionic detergent with characteristics similar to CHAPS, although it is more soluble due to a more polar head group. Useful for the solubilization of integral membrane proteins.
Soluble in water (100 mg/mL - clear, colorless to slight yellowish solution), methanol (40 mg/mL- clear, colorless solution).
Formula:
C₃₂H₅₈N₂O₈S MW: 630.89 g/mol Melting Pt: 184…186 °C (lit.) Storage Temperature: Ambient |
CAS Number:
82473-24-3 |
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
pH (10% aq soln) | 5,0 - 9,0 |
OD350 nm (1% aq soln) | <0,04 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...