Sodium cholate ≥98.5%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02102897.1 02102897.5 0210289701 0210289705 02102897.3
IC102897.1EA 204.19 USD
IC102897.1 IC102897.5 IC10289701 IC10289705 76102-630
Sodium cholate ≥98.5%, white powder
Sodium cholate

Cholic acid is a biochemical solubilizing agent (a non-ionic, non-denaturing detergent). Occurs inconjugation with glycine or taurine in bile of most vertebrates.


  • Critical Micelle Concentration (CMC): 14mM


Cholic acid is used in the solubilization of membrane-bound proteins in their native state. It is a component for the preparation of liposomes. It has also been used in a study to investigate the elastic deformable vesicles, transfersomes, for transdermal drug delivery.
Cholate is a bile acid produced in liver during cholesterol breakdown. Cholate downregulates cholesterol-7-α-hydroxylase (rate-limiting step in bile acid synthesis). Cholate, an anionic detergent, is used alone and in combination with urea to extract and reconstitute membrane proteins and protein complexes.


Cholic acid is used in the solubilization of membrane-bound proteins in their native state. It is a component for the preparation of liposomes.


Soluble in water (100 mg/ml). Soluble in ethanol (40 mg/ml - clear, colorless solution).

Formula: C₂₄H₃₉NaO₅
Melting Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00150749
CAS Number: 361-09-1
EINECS: 206-643-5

Order Now

Specification Test Results

Identity Test Passes
Purity (HPLC) ≥98.5%
Loss on Drying ≤5%
pH (2% aq soln) 7.5-9.0
Solubility (2% aq soln) Clear and Colorless
Sodium Deoxycholate ≤1.5%
Heavy Metals ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR