2-Methyl-2-propanethiol ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2-Methyl-2-propanethiol, tert-Butyl Mercaptan, tert-Butyl mercaptan, tert-Butylthiol

M0405-25ML M0405-500ML
TCM0405-025MLEA 22 USD
TCM0405-025ML TCM0405-500ML
2-Methyl-2-propanethiol ≥98.0%
2-Methyl-2-propanethiol

CAS Number: 75-66-1
MDL Number: MFCD00004857
Molecular Formula: C4H10S
Molecular Weight: 90.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Melting point (°C): -1
Flash Point (°C): -26
Specific Gravity (20/20): 0.80


Other Product Names / Synonyms: 2-Methyl-2-propanethiol

Formula: C₄H₁₀S
MW: 90.19 g/mol
Boiling Pt: 64 °C
Melting Pt: –1 °C
Density: 0.80 (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00004857
CAS Number: 75-66-1
EINECS: 200-890-2
UN: 2347
ADR: 3,II
Merck Index: 13,01578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be68907fc5fedf630cf2ca0e1fb0f18562d8b7ce&vid=be68907fc5fedf630cf2ca0e1fb0f18562d8b7ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR