3,5-Di-tert-butylbenzyl bromide ≥98.0%

Supplier: TCI America

D2653-1G D2653-5G
TCD2653-001GEA 62 USD
TCD2653-001G TCD2653-5G
3,5-Di-tert-butylbenzyl bromide ≥98.0%
3,5-Di-tert-butylbenzyl bromide

CAS Number: 62938-08-3
MDL Number: MFCD03701623
Molecular Formula: C15H23Br
Molecular Weight: 283.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Freezing point (°C): 34


Other Product Names / Synonyms: alpha-Bromo-3,5-di-tert-butyltoluene

Formula: C₁₅H₂₃Br
MDL Number: MFCD03701623
CAS Number: 62938-08-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9b4548d3226e862f03169ccca17ddac3da47bac&vid=e9b4548d3226e862f03169ccca17ddac3da47bac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR