3,3'-Diamino-N-methyldipropylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N,N-Bis(3-aminopropyl)methylamine, N-(3-Aminopropyl)-N-methyl-1,3-propanediamine

B0821-25ML B0821-500ML
TCB0821-025MLEA 26.72 USD
TCB0821-025ML TCB0821-500ML
3,3'-Diamino-N-methyldipropylamine ≥98.0% (by GC, titration analysis)
3,3'-Diamino-N-methyldipropylamine

CAS Number: 105-83-9
MDL Number: MFCD00008217
Molecular Formula: C7H19N3
Molecular Weight: 145.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 241
Flash Point (°C): 105
Specific Gravity (20/20): 0.90


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: N,N-Bis(3-aminopropyl)methylamine, N-(3-Aminopropyl)-N-methyl-1,3-propanediamine

Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 241 °C
Density: 0.90 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00008217
CAS Number: 105-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be33a831cd295ec4971bff7786360be170852e71&vid=be33a831cd295ec4971bff7786360be170852e71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR