Butyronitrile ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
n-Butyronitrile
B0768-500ML
B0768-25ML
TCB0768-500MLEA
83.93
USD
TCB0768-500ML
TCB0768-25ML
Butyronitrile ≥99.0%
Butyronitrile
CAS Number: 109-74-0
MDL Number: MFCD00001968
Molecular Formula: C4H7N
Molecular Weight: 69.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 17
Specific Gravity (20/20): 0.79
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Propyl Cyanide
Formula:
C₄H₇N MW: 69.11 g/mol Boiling Pt: 118 °C Melting Pt: -112 °C Density: 0.79 (20 °C) Flash Pt: 17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001968 CAS Number: 109-74-0 EINECS: 203-700-6 UN: 2411 ADR: 3,II Merck Index: 13,01597 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...