Butyric anhydride ≥98.0%
Supplier: TCI America
|
Danger
|
B0766-500ML
B0766-25ML
TCB0766-500MLEA
120.3
USD
TCB0766-500ML
TCB0766-25ML
Butyric anhydride ≥98.0%
Butyric anhydride
CAS Number: 106-31-0
MDL Number: MFCD00009389
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 90
Specific Gravity (20/20): 0.97
Formula:
C₈H₁₄O₃ Boiling Pt: 201 °C Melting Pt: –73 °C Density: 0.97 (20 °C) Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009389 CAS Number: 106-31-0 EINECS: 203-383-4 UN: 2739 ADR: 8,III Merck Index: 13,01594 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...