Bromoacetic acid ≥98.0%

Supplier: TCI America
Danger

B0531-25G B0531-500G
TCB0531-025GEA 31.52 USD
TCB0531-025G TCB0531-500G
Bromoacetic acid ≥98.0%
Bromoacetic acid

CAS Number: 79-08-3
MDL Number: MFCD00002678
Molecular Formula: C2H3BrO2
Molecular Weight: 138.95
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 208
Melting point (°C): 46
Flash Point (°C): 110

Formula: BrCH₂COOH
Boiling Pt: 208 °C
Melting Pt: 46 °C
Density: 1.9335 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 13,01384
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=908eb5d562f2e7f48e399a33b5eef6dd738e807e&vid=908eb5d562f2e7f48e399a33b5eef6dd738e807e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR