Pararosaniline hydrochloride C.I.42500, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride
, Basic Fuchsin Hydrochloride
, Parafuchsin hydrochloride
, Basic Red 9
, Basic Parafuchsin
, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride
, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride
, Paramagenta hydrochloride
, Pararosaniline chloride
Management of Change (MOC) category = N
S903-03
BAKRS903-03
JTS903-3CS
889.53
USD
CAJTS903-3
JTS903-3
Pararosaniline hydrochloride C.I.42500, J.T.Baker®
Pararosaniline hydrochloride
Formula:
C₁₉H₁₈ClN₃ MW: 323.82 g/mol Melting Pt: 268 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001657 CAS Number: 569-61-9 EINECS: 209-321-2 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification (by IR) | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...