Benzenesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

B0030-500G B0030-25G
TCB0030-500GEA 351 USD
TCB0030-500G TCB0030-025G
Benzenesulfonic acid ≥98.0% (by HPLC, titration analysis)
Benzenesulphonic acid

CAS Number: 26158-00-9
MDL Number: MFCD00149976
Molecular Formula: C6H6O3S
Molecular Weight: 158.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Freezing point (°C): 53

Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 50 °C
Density: 1.32 g/cm³ (47 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8,III
Merck Index: 13,01070
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b69abb87981ca08e701d6984555109306339bf0&vid=6b69abb87981ca08e701d6984555109306339bf0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR