Butyl acetate ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
A0024-500ML
A0024-25ML
TCA0024-500MLEA
31.98
USD
TCA0024-500ML
TCA0024-25ML
Butyl acetate ≥99.0%
Butyl acetate
CAS Number: 123-86-4
MDL Number: MFCD00009445
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Melting point (°C): -78
Flash Point (°C): 28
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Acetic Acid Butyl Ester
Formula:
CH₃COO(CH₂)₃CH₃ MW: 116.16 g/mol Boiling Pt: 126 °C Melting Pt: –78 °C Density: 0.88 (20 °C) Flash Pt: 28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3,III Merck Index: 13,01534 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...