The Original Nitty-Gritty® Natural Rubber-Coated Gloves, Showa
Supplier: Showa
L66NFW-08 99NFW-10 66NF-10
32891-881CS 1160.86 USD
32891-881 47727-117 47727-154 47727-152 47727-164 32891-884
The Original Nitty-Gritty® Natural Rubber-Coated Gloves, Showa
Gloves
Offer hand protection when working with sharp or abrasive materials including glass, sheet metal, scrap metal, tin plate, lumber, concrete block, and tile.
  • Natural rubber coating is cut-, puncture-, snag-, and abrasion-resistant
  • Wrinkle finish allows good wet and dry grip
Gloves feature cotton outer shell with soft, five-piece flannel inner liner for comfort. Insulated gloves feature triple-layer foam insulation sandwiched between a layer of 100% cotton jersey and 100% cotton interlock. Sanitized. Color: natural with yellow rubber coating.
Gloves enable easy handling of smooth or slippery materials and are an economical replacement for leather work gloves. They are also acceptable for use in food and pharmaceutical applications.
Certificaciones: Gloves are made using only listed materials and levels outlined in FDA 21 CFR 170-199.