Decylamine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: n-Decylamine, Decan-1-amine, 1-Aminodecane

D0033-25ML D0033-500ML
TCD0033-025MLEA 77.3 USD
TCD0033-025ML TCD0033-500ML
Decylamine ≥98.0% (by titrimetric analysis)
Decylamine

CAS Number: 2016-57-1
MDL Number: MFCD00008149
Molecular Formula: C10H23N
Molecular Weight: 157.30
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 99
Freezing point (°C): 13
Specific Gravity (20/20): 0.80


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Decylamine

Formula: H₃C(CH₂)₈CH₂NH₂
MW: 157.3 g/mol
Boiling Pt: 220 °C
Density: 0.80 (20 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00008149
CAS Number: 2016-57-1
EINECS: 217-957-7
UN: 2922
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acdba1090791775ac2b5dd72fab823ec813d2117&vid=8dfcfbe0de7eaf08c298e5a27d555f242b50cfdc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR