2-(2-Aminoethyl)-1,3-dioxolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1176-5G A1176-1G
TCA1176-005GEA 348.87 USD
TCA1176-005G TCA1176-001G
2-(2-Aminoethyl)-1,3-dioxolane ≥98.0% (by GC, titration analysis)
2-(2-Aminoethyl)-1,3-dioxolane

CAS Number: 5754-35-8
MDL Number: MFCD00142756
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 75
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C

Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Boiling Pt: 75 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00142756
CAS Number: 5754-35-8
EINECS: 227-277-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3990f2c928da51298e44e984377815d8880bf66b&vid=3990f2c928da51298e44e984377815d8880bf66b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR