trans-4-Aminocyclohexanol ≥98.0%

Supplier: TCI America

A1160-500G A1160-25G
TCA1160-500GEA 364 USD
TCA1160-500G TCA1160-25G
trans-4-Aminocyclohexanol ≥98.0%
trans-4-Aminocyclohexanol

CAS Number: 27489-62-9
MDL Number: MFCD00067698
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 110


Other Product Names / Synonyms: trans-4-Amino-1-hydroxycyclohexane

Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 127
Melting Pt: 110 °C
MDL Number: MFCD00067698
CAS Number: 27489-62-9
EINECS: 248-492-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2728d1724c07ce52f0bc46c2ab534b3c344ff0d1&vid=2728d1724c07ce52f0bc46c2ab534b3c344ff0d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR