(±)-2-Aminobutan-1-ol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A0684-25G A0684-500G
TCA0684-025GEA 22.39 USD
TCA0684-025G TCA0684-500G
(±)-2-Aminobutan-1-ol ≥98.0% (by GC, titration analysis)
(±)-2-Aminobutan-1-ol

CAS Number: 96-20-8
MDL Number: MFCD00008095
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 180
Melting point (°C): 2
Flash Point (°C): 84
Specific Gravity (20/20): 0.95

Formula: C₄H₁₁NO
Boiling Pt: 180 °C
Melting Pt: 2 °C
Density: 0.95 (20 °C)
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00008095
CAS Number: 96-20-8
EINECS: 202-488-2
UN: 2735
ADR: 8,III
Merck Index: 13,00426
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4638e76639d53b81ce5b0536bd93e942aa986da7&vid=4638e76639d53b81ce5b0536bd93e942aa986da7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR