Anisole ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Methyl phenyl ether, Methoxybenzene
A0492-500G
A0492-25G
TCA0492-500GEA
49.99
USD
TCA0492-500G
TCA0492-25G
Anisole ≥99.0%
Anisole
CAS Number: 100-66-3
MDL Number: MFCD00008354
Molecular Formula: C7H8O
Molecular Weight: 108.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Melting point (°C): -37
Flash Point (°C): 51
Specific Gravity (20/20): 1.00
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis
Other Product Names / Synonyms: Methoxybenzene, Methyl Phenyl Ether
Formula:
C₇H₈O MW: 108.14 g/mol Boiling Pt: 154 °C Melting Pt: -37 °C Density: 1.00 (20 °C) Flash Pt: 51 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008354 CAS Number: 100-66-3 EINECS: 202-876-1 UN: 2222 ADR: 3,III Merck Index: 13,00672 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...