4-Amino-3-hydroxybutyric acid ≥98.0%
Supplier: TCI America
A0318-5G
A0318-25G
TCA0318-005GEA
19
USD
TCA0318-005G
TCA0318-025G
4-Amino-3-hydroxybutyric acid ≥98.0%
4-Amino-3-hydroxybutyric acid
CAS Number: 924-49-2
MDL Number: MFCD00008141
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 218
Formula:
C₄H₉NO₃ Melting Pt: 218 °C |
MDL Number:
MFCD00008141 CAS Number: 924-49-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...