2-(4-Fluorophenoxy)-1-ethanamine 95%

Supplier: APOLLO SCIENTIFIC

PC430528-1G PC430528-250MG
77415-250EA 158.94 USD
77415-250 77415-252
2-(4-Fluorophenoxy)-1-ethanamine 95%
2-(4-Fluorophenoxy)-1-ethanamine
Formula: C₈H₁₀FNO
Storage Temperature: Ambient
MDL Number: MFCD00235183
CAS Number: 6096-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR