Heptafluorobutyryl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4490-1G PC4490-250MG
77415-444EA 33.62 USD
77415-444 77415-446
Heptafluorobutyryl chloride 97%
Heptafluorobutyryl chloride
Formula: C₄ClF₇O
Boiling Pt: 38…39 °C
Density: 1.556 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000707
CAS Number: 375-16-6
EINECS: 206-785-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR