1,2-Bis(1,1,2,2-tetrafluoroethoxy)benzene

Supplier: APOLLO SCIENTIFIC
Warning

PC4715-1G PC4715-250MG
77415-680EA 28.4 USD
77415-680 77415-682
1,2-Bis(1,1,2,2-tetrafluoroethoxy)benzene
1,2-Bis(1,1,2,2-tetrafluoroethoxy)benzene
Formula: C₁₀H₆F₈O₂
Boiling Pt: 97…100 °C/32 mmHg
Storage Temperature: Ambient
MDL Number: MFCD03424468
CAS Number: 4063-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR