2-Fluoro-5-toluoyl chloride

Supplier: APOLLO SCIENTIFIC

PC302689-250MG PC302689-500MG
77414-238EA 81 USD
77414-238 77414-240
2-Fluoro-5-toluoyl chloride
2-Fluoro-5-toluoyl chloride
Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD04973776
CAS Number: 135564-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR