4-Fluoro-m-toluonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC0774-1G PC0774-25G PC0774-5G
77412-686EA 21.26 USD
77412-686 77412-688 77412-690
4-Fluoro-m-toluonitrile 98%
4-Fluoro-m-toluonitrile
Formula: C₈H₆FN
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03788510
CAS Number: 185147-08-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR