3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3'-(Trifluoromethyl)-[1,1'-biphenyl]-2-amine hydrochloride

PC11163-1G PC11163-50MG
77413-026EA 1011.68 USD
77413-026 77413-028
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine 96%
3'-(Trifluoromethyl)[1,1'-biphenyl]-2-amine
Formula: C₁₃H₁₀F₃N
MW: 237.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424654
CAS Number: 365-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR