α-Bromopropiophenone

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Bromopropiophenone

OR907023-1G OR907023-250MG
77411-568EA 23.4 USD
77411-568 77411-570
α-Bromopropiophenone
α-Bromopropiophenone
Formula: C₆H₅C(O)CHBrCH₃
MW: 213.07 g/mol
Boiling Pt: 245…250 °C
Density: 1.417 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000145
CAS Number: 2114-00-3
EINECS: 218-307-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR