Resorcinol 99%

Supplier: APOLLO SCIENTIFIC

OR911293-100G OR911293-250G
77411-608EA 34.64 USD
77411-608 77411-610
Resorcinol 99%
Resorcinol
Formula: C₆H₆O₂
Boiling Pt: 281 °C
Melting Pt: 109…111 °C
Density: 1.272 g/cm³ (25 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002269
CAS Number: 108-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR