Methyl pyruvate 97%
Supplier: APOLLO SCIENTIFIC
OR50965-25G
OR50965-5G
77410-134EA
22.34
USD
77410-134
77410-136
Methyl pyruvate 97%
Methyl pyruvate
Formula:
C₄H₆O₃ Boiling Pt: 134…137 °C Density: 1.13 g/cm³ (25 °C) Flash Pt: 47 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008754 CAS Number: 600-22-6 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...