2-Methoxybenzylamine 98+%

Supplier: APOLLO SCIENTIFIC
Danger

OR5027-100G OR5027-25G
77410-100EA 326.02 USD
77410-100 77410-102
2-Methoxybenzylamine 98+%
2-Methoxybenzylamine
Formula: C₈H₁₁NO
Boiling Pt: 228…229 °C
Density: 1.057 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008110
CAS Number: 6850-57-3
EINECS: 229-941-7
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR