Benzylammonium chloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Benzylamine hydrochloride

OR27725-100G OR27725-25G
77407-976EA 28.28 USD
77407-976 77407-978
Benzylammonium chloride 99%
Benzylammonium chloride
Formula: C₇H₁₀ClN
Melting Pt: 262…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00012852
CAS Number: 3287-99-8
EINECS: 221-943-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR