2-Iodopropane 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR4540-1G OR4540-25G OR4540-5G
77409-854EA 68.26 USD
77409-854 77409-856 77409-858
2-Iodopropane 99%
2-Iodopropane
Formula: C₃H₇I
Boiling Pt: 88…90 °C
Melting Pt: –90 °C
Density: 1.703 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00001071
CAS Number: 75-30-9
EINECS: 200-562-9
UN: 2392
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR