1H-Pyrazolo[3,4-d]pyrimidin-4-amine 95%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR27867-10G
OR27867-1G
OR27867-5G
77408-000EA
69.28
USD
77408-000
77408-002
77408-004
1H-Pyrazolo[3,4-d]pyrimidin-4-amine 95%
1H-Pyrazolo[3,4-d]pyrimidin-4-amine
Formula:
C₅H₅N₅ Melting Pt: >325 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005688 CAS Number: 2380-63-4 EINECS: 219-174-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...