10-Undecen-1-ol 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Undecen-11-ol, 10-Undecen-1-ol

OR18341-10G OR18341-25G OR18341-50G
77406-246EA 21.26 USD
77406-246 77406-248 77406-252
10-Undecen-1-ol 99%
10-Undecen-1-ol
Formula: C₁₁H₂₂O
MW: 170.3 g/mol
Boiling Pt: 132…133 °C/15 mmHg
Melting Pt: –3 °C
Density: 0.848 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00004750
CAS Number: 112-43-6
EINECS: 203-971-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR