(R)-2-Aminopropanamide hydrochloride 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR22614-250MG
OR22614-1G
77406-914EA
21.26
USD
77406-914
77406-912
(R)-2-Aminopropanamide hydrochloride 97%
(R)-2-Aminopropanamide hydrochloride
Formula:
C₃H₉ClN₂O Melting Pt: 169…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039093 CAS Number: 71810-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...