Isoquinoline-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR16580-1G OR16580-25G OR16580-5G
77405-742EA 21.28 USD
77405-742 77405-744 77405-746
Isoquinoline-3-carboxylic acid
Isoquinoline-3-carboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075137
CAS Number: 6624-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR