tert-Butyl (piperidin-4-ylmethyl)carbamate 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (4-Piperidylmethyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Piperidylmethyl)carbamate, 4-(tert-Butoxycarbonylaminomethyl)piperidine, 4-(Boc-aminomethyl)piperidine

OR10830-25G OR10830-5G
77402-944EA 158.94 USD
77402-944 77402-946
tert-Butyl (piperidin-4-ylmethyl)carbamate 95%
tert-Butyl (piperidin-4-ylmethyl)carbamate
Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Melting Pt: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01631214
CAS Number: 135632-53-0
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR