6-Chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR111308-250MG OR111308-500MG
77403-722EA 21.26 USD
77403-722 77403-724
6-Chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylic acid
6-Chloro-2-methylimidazo[1,2-b]pyridazine-3-carboxylic acid
Formula: C₈H₆ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD08704709
CAS Number: 14714-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR