2-(Chloromethyl)-1,3-benzoxazole-5-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR01971-100MG OR01971-250MG
77401-912EA 110.04 USD
77401-912 77401-914
2-(Chloromethyl)-1,3-benzoxazole-5-carbonitrile 95%
2-(Chloromethyl)-1,3-benzoxazole-5-carbonitrile
Formula: C₉H₅ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09836175
CAS Number: 170993-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR