2-Allylbenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

OR01935-100MG OR01935-500MG
77401-906EA 172.18 USD
77401-906 77401-908
2-Allylbenzonitrile
2-Allylbenzonitrile
Formula: C₁₀H₉N
Storage Temperature: Ambient
CAS Number: 61463-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR