Tropinone oxime hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 8-Methyl-8-azabicyclo3.2.1octan-3-one oxime hydrochloride

OR018323-1G OR018323-250MG
77401-836EA 32.6 USD
77401-836 77401-838
Tropinone oxime hydrochloride 97%
Tropinone oxime hydrochloride
Formula: C₈H₁₄N₂O·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD01073570
CAS Number: 212079-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR