2-(Pyridin-4-yl)-1H-indole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(1H-Indol-2-yl)pyridine

OR01566-1G OR01566-250MG
77401-760EA 467.64 USD
77401-760 77401-762
2-(Pyridin-4-yl)-1H-indole 95%
2-(Pyridin-4-yl)-1H-indole
Formula: C₁₃H₁₀N₂
MW: 194.232 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04966830
CAS Number: 21182-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR